missing translation for 'onlineSavingsMsg'
Learn More

Iso-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10141660 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
Amber glass bottle
Metal can
Unit Size:
2.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10141660 5 L Metal can 5 L
10243850 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10141660 Supplier Fisher Chemical Supplier No. B/5000/21
  • ONLINE EXCLUSIVE PRICE
    248.00 €
    127.37 € / 5 L
    Save 120.63 €
    48% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
MDL Number 8932

Specifications

Melting Point -99°C
Color Colorless
pH 5
Boiling Point 116°C
Packaging Metal can
Quantity 5 L
Vapor Density 4
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Isobutyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.