missing translation for 'onlineSavingsMsg'
Learn More

Iso Desloratadine, TRC

Product Code. 30012320
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012320 25 mg 25mg
17941655 50 mg 50mg
17951315 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012320 Supplier Toronto Research Chemicals Supplier No. TRCI81499525MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183198-49-4
Molecular Formula C19 H19 Cl N2
Molecular Weight (g/mol) 310.82
Synonym 8-Chloro-6,11-dihydro-11-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, (11RS)-8-chloro-11-(1,2,3,6-tetrahydropyridin-4-yl)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Desloratadine Imp. B (EP)
IUPAC Name 8-chloro-11-(1,2,3,6-tetrahydropyridin-4-yl)-6,11-dihydro-5H-benzo[1,2]cyclohepta[2,4-b]pyridine
SMILES Clc1ccc2C(C3=CCNCC3)c4ncccc4CCc2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H19ClN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-7,9,12,18,21H,3-4,8,10-11H2
Formula Weight 310.1237
Percent Purity >95%
Chemical Name or Material Iso Desloratadine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.