missing translation for 'onlineSavingsMsg'
Learn More

iso-Hexane, BAKER ANALYZED, J.T. Baker™

Product Code. 15558564 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15558564 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15558564 Supplier JT Baker Supplier No. 9267.2500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Suitable for determination of Hydrocarbon Oil Index according to ISO 9377-2. Hydrocarbon oil index concentration (as RIVM oil reference standard): max. 0.5mg/L [total peaks between n-decane (C10H22) and n-tetracontane (C40H82)]

  • High quality chemical for laboratory and specialized industrial use
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Spezifikation

Melting Point 0°C
CAS 107-83-5
Color Colorless
Density 0.7g/cm3 at 20°C
Quantity 2.5 L
Flash Point −23°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula C6H14
Water Max. 0.1%
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
Boiling Point 57°C to 63°C
Solubility Information Not or partly miscible in water
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES CCCC(C)C
IUPAC Name 2-methylpentane
Molecular Weight (g/mol) 86.178
PubChem CID 7892
Formula Weight 86.18g/mol
Grade Suitable for determination of Hydrocarbon Oil Index according to ISO 9377-2
Chemical Name or Material iso-Hexane
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.