missing translation for 'onlineSavingsMsg'
Learn More

Iso-Pentane, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073365 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10121820 500 mL Amber glass bottle 500mL
10316100 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10121820 Supplier Fisher Chemical Supplier No. P/1031/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C5H12, CAS Number-78-78-4, butane, 2-methyl, 1,1-dimethylpropane, dimethylethylmethane, butanes, 1,1,2-trimethylethane, iso-pentane, iso-c5h12, isoamylhydride, ethyldimethylmethane, isopentane, 500mL, 30 deg.C, CHEBI:30362, Colorless, 72.15g/mol, QWTDNUCVQCZILF-UHFFFAOYSA-N, 2-methylbutane

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Melting Point -160°C
Color Colorless
Boiling Point 30°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 2.48
Formula Weight 72.15g/mol
Vapor Pressure 990mbar at 20°C
Physical Form Liquid
Chemical Name or Material Iso-Pentane
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylbutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.