missing translation for 'onlineSavingsMsg'
Learn More

Iso-propanol, LC/MS grade, Thermo Scientific

Product Code. 30280933 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
Product Code. Quantity unitSize
30280933 500 mL 500mL
30280934 1 L 1 L
30280935 2.5 L 2.5 L
3 options
This item is not returnable. View return policy

Product Code. 30280933

Brand: Thermo Scientific T004610500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 60.1 g/mol

Specifications

Boiling Point 81°C
Quantity 500 mL
CAS 67-63-0
Packaging Glass Bottle
Percent Purity 99.90%
Grade LC-MS
Chemical Name or Material Isopropanol
compliance-icons compliance-icons
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.