missing translation for 'onlineSavingsMsg'
Learn More

Iso-propanol, LC/MS grade, Thermo Scientific
GREENER_CHOICE

Product Code. 30280934 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280934 1 L 1 L
30280933 500 mL 500mL
30280935 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280934 Supplier Thermo Scientific Supplier No. T004611000
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    84.80 € / 1 L
    Save 30.20 €
    26% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 60.1 g/mol

Specifications

Quantity 1 L
CAS 67-63-0
Packaging Glass Bottle
Boiling Point 81°C
Percent Purity 99.90%
Grade LC-MS
Chemical Name or Material Isopropanol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.