missing translation for 'onlineSavingsMsg'
Learn More

Iso-Propyl Acetate, Pure, Fisher Chemical™

Product Code. 10792544 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792544 2.5 L Plastic Coated Glass bottle 2.5 L
10366190 500 mL Amber glass bottle 500mL
10509200 2.5 L Amber glass bottle 2.5 L
12696757 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10792544 Supplier Fisher Chemical Supplier No. P/7640/PB17
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number 8877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Melting Point -73°C
Color Colorless
Boiling Point 88.8°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Vapor Density &3.5
Formula Weight 102.13g/mol
Vapor Pressure 61mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • iso-Propyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.