missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Propyl Alcohol-OD, CDN
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Isopropanol-d |
| CAS | 3979-51-9 |
| Molecular Formula | C3 D H7 O |
| Quantity | 25 g |
| Synonym | iso-Propyl Alcohol-OD, 2-Propanol-d (9 CI), Isopropyl alcohol-d (6 CI, 7 CI, 8 CI), 2-Deuteriooxypropane, 2-Propanol-O-d1, i-Propanol-d, Isopropanol-d |
| InChI Formula | InChI=1 S/C3H8O/c1-3(2)4/h3-4 H,1-2H3/i4D |
| SMILES | [2 H]OC(C)C |
| IUPAC Name | 2-deuteriooxypropane |
| Molecular Weight (g/mol) | 61.1 |
| Formula Weight | 61.0638 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction