missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Safety Sign Roll without Text - Warning: Biological Hazard

Product Code. 15956475 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
43 x 50 mm
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15956475 43 x 50 mm Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15956475 Supplier Brady™ Supplier No. 827648
  • ONLINE EXCLUSIVE PRICE
    167.00 €
    158.75 € / Pack of 250
    Save 8.25 €
    4% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safety sign for hazard identification.

  • Sign compliant to ISO 7010
  • Improves workplace safety

Specifications

Alternate Part Number PIC W009- Tri 050-PE-ROLL/1
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 43 x 50 mm
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.