missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Safety Sign Roll without Text - Warning: Hot Surface

Product Code. 16694352
Change view
Click to view available options
Dimensions (L x W):
87 x 100 mm
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
16694352 87 x 100 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16694352 Supplier Brady™ Supplier No. 828834
  • ONLINE EXCLUSIVE PRICE
    433.00 €
    415.72 € / 1 rolls
    Save 17.28 €
    3% Off
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Safety sign for hazard identification.

  • Sign compliant to ISO 7010
  • Improves workplace safety

Specifications

Alternate Part Number PIC W017-Tri 100-PE-ROLL/1
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 87 x 100 mm
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.