Learn More
Brady™ ISO Safety Sign without Text - Do Not Wear Gloves
Product Code. 16170541
Change view
Click to view available options
Material:
Laminated Polyester
Unit Size:
Pack of 2
Product Code. Material unitSize
16170541 Laminated Polyester Pack of 2
1 options
Product Code. 16170541 Supplier Brady™ Supplier No. 825570
  • ONLINE EXCLUSIVE PRICE
    5.85 €
    5.60 € / Pack of 2
    Save 0.25 €
    4% Off
In Stock
Add to basket
Add to basket
Safety sign for indicating mandatory action.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC P028-Dia 050-PE-SHEET/2
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Diameter (Metric) 50 mm
Shape Round
Color Red on White