missing translation for 'onlineSavingsMsg'
Learn More

iso-Valeraldehyde, TRC

Product Code. 30453574
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453574 100 mL 100mL
30449057 25 mL 25mL
30452125 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453574 Supplier Toronto Research Chemicals Supplier No. TRCV091300100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-86-3
Molecular Formula C5 H10 O
Molecular Weight (g/mol) 86.13
Synonym Butanal, 3-methyl-, Butyraldehyde, 3-methyl- (7CI), Isovaleraldehyde (8CI), 3-Methylbutanal, 3-Methyl-1-butanal, 3-Methylbutyraldehyde, Isoamylaldehyde, Isopentanal, Isovaleral, Isovaleric aldehyde, NSC 404119, iso-Valeraldehyde, β-Methylbutanal
IUPAC Name 3-methylbutanal
SMILES CC(C)CC=O

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C5H10O/c1-5(2)3-4-6/h4-5H,3H2,1-2H3
Formula Weight 86.0732
Chemical Name or Material 3-Methylbutyraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.