missing translation for 'onlineSavingsMsg'
Learn More

Isoalantolactone, MedChemExpress

Product Code. 16464887
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464887 50 mg 50mg
16449098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16464887 Supplier Medchem Express Supplier No. HYN0780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isoalantolactone is an apoptosis inducer, which also acts as an alkylating agent.

Isoalantolactone is an apoptosis inducer, which also acts as an alkylating agent.

Specifications

Chemical Name or Material Isoalantolactone
CAS 470-17-7
Color White
Purity Grade Notes Research
Molecular Formula C15H20O2
Quantity 50 mg
Synonym (+)-Isoalantolactone Isohelenin
Solubility Information DMSO : 100 mg/mL (430.44 mM; Need ultrasonic)
SMILES O=C(O[C@@](C[C@@]1(C)CCC2)([H])[C@]3([H])C[C@@]1([H])C2=C)C3=C
Molecular Weight (g/mol) 232.32
Formula Weight 232.32
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.