missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl nitrite, 97%, stabilized

Product Code. 10782201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782201 500 mL Glass Bottle 500mL
10675602 100 mL Glass Bottle 100mL
10669293 2.5 L Plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782201 Supplier Thermo Scientific Acros Supplier No. 165705000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-46-3 , 497-19-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00002057
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
PubChem CID 8053
ChEBI CHEBI:2691
IUPAC Name 3-methylbutyl nitrite
SMILES CC(C)CCON=O

Specifications

Color Yellow
Density 0.8700g/mL
Boiling Point 96.0°C to 99.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH2ONO
Refractive Index 1.3840 to 1.3880
Quantity 500 mL
Beilstein 01, 402
Fieser 01,40; 04,270; 05,358; 06,307; 09,258
Merck Index 15, 5168
Specific Gravity 0.87
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: miscible with alcohol,chloroform and ether
Stabilizer 0.2% sodium carbonate
Formula Weight 117.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isoamyl nitrite, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isoamyl nitrite, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.