missing translation for 'onlineSavingsMsg'
Learn More

Isobutanesulfonyl chloride, 97%

Product Code. 10714872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10714872 1 g Glass bottle 1g
10129689 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10714872 Supplier Thermo Scientific Acros Supplier No. 438410010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35432-36-1 | C4H9ClO2S | 156.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35432-36-1
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.63
InChI Key NQLNHTOCFWUYQE-UHFFFAOYSA-N
Synonym isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
PubChem CID 182236
IUPAC Name 2-methylpropane-1-sulfonyl chloride
SMILES CC(C)CS(=O)(=O)Cl

Specifications

Color Brown to Colorless
Boiling Point 77°C to 78°C (11.0 Torr)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 156.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isobutanesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Isobutanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.