missing translation for 'onlineSavingsMsg'
Learn More

Isobutanol, HPLC Grade, 99+%

Product Code. 11388147
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
missing translation for 'unitSize'
1 L
4 L
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11388147

missing translation for 'mfr': Thermo Scientific Alfa Aesar 022908.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05% | CAS: 78-83-1 | C4H10O | 74.123 g/mol

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Especificaciones

Melting Point -108°C
Color Colorless
Density 0.802 g/cm3
Boiling Point 107°C to 108°C
Flash Point 28°C (82°F)
Ignition Point 390°C (734°F)
Assay Percent Range >99%
Odor Sweet
Refractive Index 1.396
Quantity 1 L
UN Number 1212
Beilstein 1730878
Solubility Information Miscible with alcohol and diethyl ether. Partially miscible with water.
Formula Weight 74.12
Vapor Pressure 4-40 hPa (3-30mm Hg) at 20°C
Grade HPLC
Physical Form Liquid
Chemical Name or Material Isobutanol
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Isobutanol
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contains : Isobutanol

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado