missing translation for 'onlineSavingsMsg'
Learn More

Isobutanol, HPLC Grade, 99+%

Product Code. 11398147
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398147 4 x 1 L 4 L
11388147 1 L 1 L
11308157 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398147 Supplier Thermo Scientific Alfa Aesar Supplier No. 022908.M1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05% | CAS: 78-83-1 | C4H10O | 74.123 g/mol

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point °C to 108°C
Color Colorless
Density 0.802
Boiling Point 107°C to 108°C
Flash Point 28°C (82°F)
Ignition Point 390°C (734°F)
Assay Percent Range >99%
Odor Sweet
Refractive Index 1.396
Quantity 4 x 1 L
Beilstein 1730878
Solubility Information Miscible with alcohol and diethyl ether. Partially miscible with water.
Formula Weight 74.12
Vapor Pressure 4-40 hPa (3-30mm Hg) at 20°C
Grade HPLC
Physical Form Liquid
Chemical Name or Material Isobutanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.