missing translation for 'onlineSavingsMsg'
Learn More

Isobutanol, Supelco™

Product Code. 15824000
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824000 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15824000 Supplier Supelco Supplier No. 1009841000
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For analysis EMSURE™ ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutanol
Melting Point -108°C
CAS 78-83-1
Density 0.802 g/cm³ at 20°C
Quantity 1 L
Flash Point 28°C (closed cup)
Molecular Formula (CH3)2CHCH2OH
Synonym Isobutanol, Isobutylalcohol, 2-Methyl-1-propanol, Iso-Butanol, Isobutanol
Boiling Point 106°C to 108°C (1013 hPa)
Molecular Weight (g/mol) 74.12
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.