missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15578514 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578514 4 L Narrow Mouth Amber Glass Bottle 4 L
15568514 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15578514 Supplier Avantor J.T.Baker Supplier No. 904403
  • ONLINE EXCLUSIVE PRICE
    407.90 €
    337.47 € / 4 L
    Save 70.43 €
    17% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol

Specifications

CAS 78-83-1
Flash Point 28.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4 L
Specific Gravity 1L = 0.80kg
Molecular Weight (g/mol) 74.12
Percent Purity ≥99.0%
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Liquid
Chemical Name or Material Isobutanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.