missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl isobutyrate, 98%

Product Code. 11391647
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 11391647

Brand: Thermo Scientific Alfa Aesar L04655.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-85-8 | C8H16O2 | 144.21 g/mol

Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

Solubility
Soluble in water (1 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-85-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00008916
InChI Key RXGUIWHIADMCFC-UHFFFAOYSA-N
Synonym isobutyl isobutyrate, isobutyl 2-methylpropanoate, isobutyl isobutanoate, isobutyric acid, isobutyl ester, propanoic acid, 2-methyl-, 2-methylpropyl ester, 2-methylpropyl 2-methylpropionate, 2-methylpropyl isobutyrate, isobutylester kyseliny isomaselne, fema number 2189, 2-methyl-1-propyl 2-methylpropanoate
PubChem CID 7351
IUPAC Name 2-methylpropyl 2-methylpropanoate
SMILES CC(C)COC(=O)C(C)C

Specifications

Melting Point -81°C
Boiling Point 147°C to 149°C
Flash Point 37°C (98°F)
Odor Fruit-like
Quantity 250 mL
UN Number UN2528
Beilstein 1701355
Merck Index 14,5145
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material Isobutyl isobutyrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyl isobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Isobutyl isobutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.