missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10069571
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10069571 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10069571 Supplier Thermo Scientific Maybridge Supplier No. SB01703EA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 591-82-2
CAS Min % 97.0
Density 0.9400g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H9NS
Linear Formula (CH3)2CHCH2NCS
MDL Number MFCD00022062
Quantity 10 g
Synonym isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
InChI Key NSDDRJXKROCWRZ-UHFFFAOYSA-N
SMILES CC(C)CN=C=S
IUPAC Name 1-isothiocyanato-2-methylpropane
Molecular Weight (g/mol) 115.194
PubChem CID 68960
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material Isobutyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.