missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl isothiocyanate, 97%

Product Code. 11423178
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423178 5 g 5g
11433178 25 g 25g
11443178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423178 Supplier Thermo Scientific Alfa Aesar Supplier No. A18932.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 591-82-2 | C5H9NS | 115.194 g/mol

It is isothiocyanate compound used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is isothiocyanate compound used for proteomics research.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-82-2
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.194
MDL Number MFCD00022062
InChI Key NSDDRJXKROCWRZ-UHFFFAOYSA-N
Synonym isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
PubChem CID 68960
IUPAC Name 1-isothiocyanato-2-methylpropane
SMILES CC(C)CN=C=S

Spécification

Density 0.94
Boiling Point 160°C to 162°C
Flash Point 24°C (75°F)
Odor Pungent
Refractive Index 1.498
Quantity 5 g
UN Number UN2924
Beilstein 1740371
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material Isobutyl isothiocyanate
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.