Learn More
Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol
Description
Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.889 g/mL |
| Boiling Point | 155°C |
| Flash Point | 46°C (115°F) |
| Quantity | 4 L |
| UN Number | UN2283 |
| Beilstein | 1747595 |
| Solubility Information | Soluble in water at 20°C,2g/L |
| Formula Weight | 142.2 |
| Percent Purity | ≥99.5% |
| Physical Form | Liquid |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.