missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol

Product Code. 11384927
Click to view available options
Quantity:
250 mL
1 L
4 L
Unit Size:
1 L
250mL
4 L
This item is not returnable. View return policy

Product Code. 11384927

Brand: Thermo Scientific Alfa Aesar 041948.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-86-9 | C8H14O2 | 142.20 g/mol

Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-86-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00008931,MFCD00084435
InChI Key RUMACXVDVNRZJZ-UHFFFAOYSA-N
Synonym isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
PubChem CID 7352
IUPAC Name 2-methylpropyl 2-methylprop-2-enoate
SMILES CC(C)COC(=O)C(C)=C

Specifications

Density 0.889 g/mL
Boiling Point 155°C
Flash Point 46°C (115°F)
Quantity 1 L
UN Number UN2283
Beilstein 1747595
Solubility Information Soluble in water at 20°C,2g/L
Formula Weight 142.2
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Isobutyl methacrylate, Stabilized with 10ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.