missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Nitrite, TRC

Product Code. 30009490
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009490 500 mg 500mg
30005457 1 g 1g
30004652 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009490 Supplier Toronto Research Chemicals Supplier No. TRCI357945500MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-56-3
Molecular Formula C4 H9 N O2
Molecular Weight (g/mol) 103.12
Synonym Nitrous acid, isobutyl ester, 2-Methylpropyl Nitrite, Blackjack, iso-Butyl Nitrite;
IUPAC Name 2-methylpropyl nitrite
SMILES CC(C)CON=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H9NO2/c1-4(2)3-7-5-6/h4H,3H2,1-2H3
Formula Weight 103.063
Chemical Name or Material Isobutyl Nitrite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.