missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine

Product Code. 10071514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071514 250 mL Glass bottle 250mL
10366694 1 L Glass bottle 1 L
10420734 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071514 Supplier Thermo Scientific Acros Supplier No. 122502500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-81-9 | C4H11N | 73.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-81-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008146
InChI Key KDSNLYIMUZNERS-UHFFFAOYSA-N
Synonym isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
PubChem CID 6558
ChEBI CHEBI:15997
IUPAC Name 2-methylpropan-1-amine
SMILES CC(C)CN

Specifications

Melting Point -85.0°C
Color Colorless to Yellow
Density 0.7300g/mL
Boiling Point 64.0°C to 71.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2CHCH2NH2
Refractive Index 1.397
Quantity 250 mL
Beilstein 04, 163
Merck Index 15, 5177
Specific Gravity 0.73
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 73.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Isobutylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.