missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene oxide, 98%

Product Code. 10653512 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10653512 100 mL Glass bottle 100mL
10707392 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10653512 Supplier Thermo Scientific Supplier No. 348501000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 558-30-5 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 558-30-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00066354
InChI Key GELKGHVAFRCJNA-UHFFFAOYSA-N
Synonym isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
PubChem CID 11208
IUPAC Name 2,2-dimethyloxirane
SMILES CC1(CO1)C

Spezifikation

Color Colorless
Density 0.8080g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3710 to 1.3730
Quantity 100 mL
Beilstein 17, V,1, 58
Specific Gravity 0.808
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether
Viscosity 0.513 mm2/s (20°C)
Formula Weight 72.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutylene oxide, 98%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Isobutylene oxide
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 C
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.