missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.9410g/mL |
| Boiling Point | 34.6°C |
| Flash Point | −34°C |
| Packaging | AcroSeal™ Glass bottle |
| Linear Formula | (CH3)2CHCH2MgBr |
| Quantity | 800 mL |
| Specific Gravity | 0.941 |
| Formula Weight | 161.32 |
| Chemical Name or Material | Isobutylmagnesium bromide, 2M solution in diethyl ether |
Safety and Handling
Product Identifier
- Isobutylmagnesium bromide, 2M solution in diethyl ether
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 2
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H261-In contact with water releases flammable gases.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction