missing translation for 'onlineSavingsMsg'
Learn More

Isobutyltrimethoxysilane, 97%

Product Code. 11490137
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490137 25 g 25g
11400147 100 g 100g
11410147 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490137 Supplier Thermo Scientific Alfa Aesar Supplier No. B21048.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18395-30-7 | C7H18O3Si | 178.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18395-30-7
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.303
MDL Number MFCD00051589
InChI Key XYJRNCYWTVGEEG-UHFFFAOYSA-N
Synonym isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane
PubChem CID 87622
IUPAC Name trimethoxy(2-methylpropyl)silane
SMILES CC(C)C[Si](OC)(OC)OC

Specifications

Boiling Point 154°C to 157°C
Quantity 25 g
Flash Point 43°C (109°F)
Refractive Index 1.396
Sensitivity Moisture sensitive
Density 0.93
UN Number UN1993
Beilstein 2233976
Formula Weight 178.31
Percent Purity 97%
Chemical Name or Material Isobutyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.