missing translation for 'onlineSavingsMsg'
Learn More

Isobutyramidoxime, 96%

Product Code. 11499590
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499590 250 mg 250mg
11409600 1 g 1g
15407897 5 g 5g
15417897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11499590 Supplier Thermo Scientific Alfa Aesar Supplier No. H50892.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35613-84-4 | C4H10N2O | 102.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35613-84-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD03426251,MFCD08705910,MFCD00464225
InChI Key JHRDEHLFNLLCQS-UHFFFAOYSA-N
Synonym z-n'-hydroxyisobutyrimidamide, 1z-n'-hydroxy-2-methylpropanimidamide, n-hydroxyisobutyrimidamide, n-hydroxy-2-methylpropanimidamide, z-n'-hydroxy-2-methylpropimidamide, n-hydroxyisobutyramidine, z-n'-hydroxy-2-methylpropanimidamide, isobutyramidoxime 250mg, n/'-hydroxy-2-methylpropanimidamide, propanimidamide, n-hydroxy-2-methyl
PubChem CID 9582835
IUPAC Name N'-hydroxy-2-methylpropanimidamide
SMILES CC(C)C(\N)=N/O

Specifications

Melting Point 57°C to 65°C
Quantity 250 mg
Formula Weight 102.14
Percent Purity 96%
Chemical Name or Material Isobutyramidoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.