missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric anhydride, 97%

Product Code. 10411291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Glass bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411291 500 g Glass bottle 500g
10277590 2 kg Glass bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411291 Supplier Thermo Scientific Acros Supplier No. 257785000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-72-3 | C8H14O3 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-72-3 , 79-09-4 , 64-19-7
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
MDL Number MFCD00008913
InChI Key LSACYLWPPQLVSM-UHFFFAOYSA-N
Synonym isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
PubChem CID 7346
ChEBI CHEBI:84261
IUPAC Name 2-methylpropanoyl 2-methylpropanoate
SMILES CC(C)C(=O)OC(=O)C(C)C

Specifications

Melting Point -53.0°C
Color Colorless
Density 0.9540g/mL
Boiling Point 182.0°C
Flash Point 66.4°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)CHCO]2O
Refractive Index 1.4050 to 1.4070
Quantity 500 g
Beilstein 02, 292
Specific Gravity 0.954
Solubility Information Solubility in water: decomposes
Viscosity 1.18 mPa.s (25°C)
Formula Weight 158.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isobutyric anhydride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyric anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.