missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Anhydride, TRC

Product Code. 30404706
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404706 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404706 Supplier Toronto Research Chemicals Supplier No. TRCI789190500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutyric Anhydride
CAS 97-72-3
Molecular Formula C8H14O3
Quantity 500 mL
Synonym 2-Methylpropanoic Acid 1,1'-Anhydride, 2-Methylpropanoic Acid Anhydride, 2-Methylpropanoic Anhydride, 2-Methylpropionic Acid Anhydride, 2-Methylpropionic Anhydride, Isobutanoic Anhydride, Isobutyric Acid Anhydride, Isobutyryl Anhydride
InChI Formula InChI=1S/C8H14O3/c1-5(2)7(9)11-8(10)6(3)4/h5-6H,1-4H3
SMILES CC(C)C(=O)OC(=O)C(C)C
IUPAC Name 2-methylpropanoyl 2-methylpropanoate
Molecular Weight (g/mol) 158.19
Formula Weight 158.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.