missing translation for 'onlineSavingsMsg'
Learn More

Isobutyrophenone 97%, ACROS Organics™

Product Code. 10071680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071680 25 g Glass bottle 25g
10295770 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071680 Supplier Thermo Scientific Acros Supplier No. 157110250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 611-70-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008917
InChI Key BSMGLVDZZMBWQB-UHFFFAOYSA-N
Synonym isobutyrophenone, isopropyl phenyl ketone, 2-methylpropiophenone, 1-propanone, 2-methyl-1-phenyl, phenyl isopropyl ketone, isopropyl phenyl keton, alpha-methylpropiophenone, unii-8l53972ntz, .alpha.-methylpropiophenone, 2-methyl-1-phenyl-1-propanone
PubChem CID 69144
IUPAC Name 2-methyl-1-phenylpropan-1-one
SMILES CC(C)C(=O)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 0.9880g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCH(CH3)2
Refractive Index 1.5162 to 1.5182 (20°C, 589nm)
Quantity 25 g
Beilstein 07, 316
Formula Weight 148.2
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Isobutyrophenone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.