missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl chloride, 98%

Product Code. 10255730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255730 250 mL Glass bottle 250mL
10583521 10 mL Glass bottle 10mL
10275680 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10255730 Supplier Thermo Scientific Acros Supplier No. 160662500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 79-30-1 | C4H7ClO | 106.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-30-1
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000717
InChI Key DGMOBVGABMBZSB-UHFFFAOYSA-N
Synonym isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
PubChem CID 62325
IUPAC Name 2-methylpropanoyl chloride
SMILES CC(C)C(=O)Cl

Specifications

Melting Point -90.0°C
Density 1.0170g/mL
Boiling Point 91.0°C to 93.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCl
Refractive Index 1.4060 to 1.4080
Quantity 250 mL
Beilstein 02, 293
Specific Gravity 1.017
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether
Viscosity 0.55 mPa.s (20°C)
Formula Weight 106.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyryl chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.