missing translation for 'onlineSavingsMsg'
Learn More

Isochlortetracycline, TRC

Product Code. 30011885
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011885 5 mg 5mg
30009542 10 mg 10mg
30010004 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011885 Supplier Toronto Research Chemicals Supplier No. TRCI7900005MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514-53-4
Molecular Formula C22 H23 Cl N2 O8
Molecular Weight (g/mol) 478.88
Synonym 2-Naphthalenecarboxamide, 6-[(1S)-7-chloro-1,3-dihydro-4-hydroxy-1-methyl-3-oxo-1-isobenzofuranyl]-4-(dimethylamino)-1,4,4a,5,6,7,8,8a-octahydro-3,8a-dihydroxy-1,8-dioxo-, (4S,4aS,6S,8aS)-, 2-Naphthalenecarboxamide, 6-(7-chloro-1,3-dihydro-4-hydroxy-1-methyl-3-oxo-1-isobenzofuranyl)-4-(dimethylamino)-1,4,4a,5,6,7,8,8a-octahydro-3,8a-dihydroxy-1,8-dioxo-, [4S-[4α,4aα,6β(R*),8aα]]-, Phthalide, 3-[6-carbamoyl-8-(dimethylamino)-1,2,3,4,4a,5,8,8a-octahydro-4a,7-dihydroxy-4,5-dioxo-2-naphthyl]-4-chloro-7-hydroxy-3-methyl- (6CI,7CI,8CI), (4S,4aS,6S,8aS)-6-[(1S)-7-Chloro-1,3-dihydro-4-hydroxy-1-methyl-3-oxo-1-isobenzofuranyl]-4-(dimethylamino)-1,4,4a,5,6,7,8,8a-octahydro-3,8a-dihydroxy-1,8-dioxo-2-naphthalenecarboxamide, 7-Chloroisotetracycline, Iso-chlorotetracycline, Isoaureomycin, Isochlortetracycline, (4S,4aS,6S,8aS)-6-[(1S)-7-Chloro-4-hydroxy-1-methyl-3-oxo-isobenzofuran-1-yl]-4-(dimethylamino)-3,8a-dihydroxy-1,8-dioxo-4a,5,6,7-tetrahydro-4H-naphthalene-2-carboxamide
IUPAC Name (4S,4aS,6S,8aS)-6-[(1S)-7-chloro-4-hydroxy-1-methyl-3-oxo-2-benzofuran-1-yl]-4-(dimethylamino)-3,8a-dihydroxy-1,8-dioxo-4a,5,6,7-tetrahydro-4H-naphthalene-2-carboxamide
SMILES CN(C)[C@H]1[C@@H]2C[C@@H](CC(=O)[C@]2(O)C(=O)C(=C1O)C(=O)N)[C@]3(C)OC(=O)c4c(O)ccc(Cl)c34

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H23ClN2O8/c1-21(15-10(23)4-5-11(26)13(15)20(31)33-21)8-6-9-16(25(2)3)17(28)14(19(24)30)18(29)22(9,32)12(27)7-8/h4-5,8-9,16,26,28,32H,6-7H2,1-3H3,(H2,24,30)/t8-,9-,16-,21-,22-/m0/s1
Formula Weight 478.1143
Chemical Name or Material Isochlortetracycline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.