Learn More
Isofield™ Isoclave CF Autoclavable Cleanroom Goggles - Grade A/B Compatible
BIOPHARMA
Product Code. 30178822
Change view
Click to view available options
Frame Material:
Silicone
Unit Size:
Pack of 50
Product Code. Frame Material Unit Size
30178822 Silicone Pack of 50
1 options
Product Code. 30178822 Supplier Isofield™ Supplier No. 50210
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Autoclavable goggle with top mounted indirect venting. Made with soft silicon housing to ensure comfort, and a wide view anti-fog coated lens. Suitable for autoclaving at 121°C for 30 min., the Isoclave CF offers up to 35 cycles on the lens and 40 on straps and housing, under lab conditions.
  • Close-fit autoclavable cleanroom goggles
  • Anti-fog
  • Indirect upper ventilation
  • Non-vented base for added Cleanroom compliance
  • Panoramic visibility
  • Fits over standard prescription glasses
  • Ergonomic frame with flexible nose skirt to ensure secure fit
  • Premium anti-scratch
  • Non-sterile - goggle must be processed prior to use
Specifications
Product Type Cleanroom Goggles
Disposable Single-use
Lens Material Polycarbonate (Medical Grade)
Frame Material Silicone