Learn More
Isofield™ Isoclave IV Autoclavable Cleanroom Goggles - Grade A/B Compatible
BIOPHARMA
Product Code. 30178836
Change view
Click to view available options
Frame Material:
Thermoplastic Rubber (TPR)
Unit Size:
Pack of 50
Product Code. Frame Material Unit Size
30178836 Thermoplastic Rubber (TPR) Pack of 50
1 options
Product Code. 30178836 Supplier Isofield™ Supplier No. 50110
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Autoclavable goggle with indirect venting. Made with a super soft thermoplastic housing to ensure comfort, and a wide view anti-fog coated lens. Suitable for autoclaving at 121°C for 30 min., the Isoclave IV offers up to 35 cycles on the lens and 40 on straps and housing, under lab conditions.
  • Indirect upper and lower ventilation
  • Panoramic visibility
  • Fits over standard prescription glasses
  • Ergonomic frame with flexible nose skirt
  • Premium anti-scratch, optical grade, anti-fog lens
  • Tested against EN 166 and ISO 16321-1
  • Certified to PPE Cat II
  • Non-sterile - Goggle must be processed prior to use
Specifications
Product Type Cleanroom Goggles
Disposable Single-use
Lens Material Polycarbonate (Medical Grade)
Frame Material Thermoplastic Rubber (TPR)