missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Boot Covers
BIOPHARMA

Product Code. 15694904 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15694904 Small Pack of 100
15604914 Medium Pack of 100
15614914 Large Pack of 100
15624914 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15694904 Supplier DuPont™ Supplier No. D15466072
Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. Dupont™ Tyvek™ IsoClean™ Boot Covers provide an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting

  • Available in sizes S to XL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunnelled elastication at shin and ankles.
  • Ankle ties.
  • Gripper™ soles provide enhanced slip-retardance and robustness.
TRUSTED_SUSTAINABILITY

Specifications

Size Small
Material Tyvek™ IsoClean™
Color White
Disposable Single-use
Sterility Sterile
Sole Type Anti-skid
Seam Type Bound
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics, Other Critical Environments
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
Packaging Quantity 100/Cs.
Packaging Type Individually Packaged in Pairs
Product Type Controlled Environments Boot Covers
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.