missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isodecyl Diphenyl Phosphate-d10, TRC
Specifications
Specifications
| Chemical Name or Material | Isodecyl Diphenyl Phosphate-d10 |
| Molecular Formula | C22 D10 H21 O4 P |
| Quantity | 25 mg |
| Synonym | Diacizer D 148-d10, Diphenyl Isodecyl Phosphate-d10, Isodecyl Alcohol Diphenyl Phosphate-d10, Phosflex 390-d10, Santicizer 2148-d10, Santicizer C 148-d10 |
| InChI Formula | InChI=1S/C22H31O4P/c1-20(2)14-8-4-3-5-13-19-24-27(23,25-21-15-9-6-10-16-21)26-22-17-11-7-12-18-22/h6-7,9-12,15-18,20H,3-5,8,13-14,19H2,1-2H3/i6D,7D,9D,10D,11D,12D,15D,16D,17D,18D |
| SMILES | [2H]c1c([2H])c([2H])c(OP(=O)(OCCCCCCCC(C)C)Oc2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H] |
| IUPAC Name | 8-methylnonyl bis(2,3,4,5,6-pentadeuteriophenyl) phosphate |
| Molecular Weight (g/mol) | 400.514 |
| Formula Weight | 400.259 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?