missing translation for 'onlineSavingsMsg'
Learn More

Isododecane, mixture of isomers, tech. 80%

Product Code. 11402357
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
Product Code. Quantity unitSize
11402357 2500 mL 2500mL
11492347 500 mL 500mL
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11402357

Brand: Thermo Scientific Alfa Aesar A14235.0F

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 31807-55-3 | C12H26 | 170.34 g/mol

Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31807-55-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00036106
InChI Key GTJOHISYCKPIMT-UHFFFAOYSA-N
Synonym isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers
PubChem CID 23459
IUPAC Name 2-methylundecane
SMILES CCCCCCCCCC(C)C

Specifications

Density 0.74
Boiling Point 170°C to 195°C
Flash Point 42°C (108°F)
Assay Percent Range 80%
Refractive Index 1.421
Quantity 2500 mL
UN Number UN3295
Solubility Information Immiscible with water.
Formula Weight 170.34
Percent Purity 80%
Grade Technical
Physical Form Liquid
Chemical Name or Material Isododecane, mixture of isomers
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isododecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Isododecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.