missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Isoguvacine hydrochloride

Product Code. 18065822
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065822 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18065822 Supplier R&D Systems™ Supplier No. 0235/100
  • ONLINE EXCLUSIVE PRICE
    312.05 €
    280.35 € / 100mg
    Save 31.70 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective GABAA agonist

Specific GABAA receptor agonist. Suppresses low magnesium induced seizure like events in organotypic hippocampal brain slices.
TRUSTED_SUSTAINABILITY

Specifications

CAS 68547-97-7
Quantity 100 mg
Product Line Tocris
Molecular Formula C6H10ClNO2
Formula Weight 163.6
Purity >99%
Molecular Weight (g/mol) 163.601
Chemical Name or Material Isoguvacine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.