missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline hydrochloride, 95%, Thermo Scientific™

Product Code. 15419448
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419448 250 mg 250mg
15499438 1 g 1g
15409448 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419448 Supplier Thermo Scientific Alfa Aesar Supplier No. H63012.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoindoline hydrochloride is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inder gas. Store away aeay from oxidizing agents, water/ moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
IUPAC Name 2,3-dihydro-1H-isoindole;hydrochloride
SMILES Cl.C1NCC2=CC=CC=C12

Specifications

Melting Point 255°C to 256°C
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 155.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Isoindoline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.