Learn More
SymLine Isolating Amplifier
Product Code. 15922531
Change view
Click to view available options
For Use With (Equipment):
Signalbox T1, T5, Signalbox2
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15922531 Signalbox T1, T5, Signalbox2 Each
1 options
Product Code. 15922531 Supplier SymLine Supplier No. 108278
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    309.25 € / Pack of 1
    Save 51.75 €
    14% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Switch amplifier for Signalbox devices.
  • Suitable for use with Signalbox T1, T5, Signalbox2
Specifications
Description Switch amplifier EU, 2 channels safety barrier, 120 - 230 V AC supply, contact or NAMUR-inputs / ATEX: Ex II (1) G [Ex ia] IIC, Ex II (1) D [Ex ia] IIIC
For Use With (Equipment) Signalbox T1, T5, Signalbox2
Product Type Switch Amplifier