missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Isoliquiritigenin

Product Code. 16408868
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408868 Supplier MedChemExpress Supplier No. HYN0102
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isoliquiritigenin is an anti-tumor flavonoid from the root of Glycyrrhiza glabra, which inhibits aldose reductase with an IC50 of 320 nM. Isoliquiritigenin is a potent inhibitor of influenza virus replication with an EC50 of 24.7 μM.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 256.25 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 961-29-5
Product Type Natural Products
Purity 98.17%
Solubility Ethanol : 100 mg/mL (390.24 mM∣Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (390.24 mM)
Synonym GU17∣ ISL∣ Isoliquiritigen∣ SJ000286237
Target Aldose Reductase∣Influenza Virus∣Autophagy∣Apoptosis
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.