missing translation for 'onlineSavingsMsg'
Learn More

Isomalt, TRC

Product Code. 30137027
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137027 250 mg 250mg
17998114 25 mg 25mg
17960095 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137027 Supplier Toronto Research Chemicals Supplier No. TRCI821065250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64519-82-0
Molecular Formula C12 H24 O11
Molecular Weight (g/mol) 344.31
Synonym Isomalt, (2ξ)-6-O-α-D-Glucopyranosyl-D-arabino-hexitol, C-Isomaltidex, GalenIQ 801, GalenIQ 980, Hydrogenated isomaltulose, IsoMaltidex 16502, Isomalt, Isomalt DC 110, Isomalt PF, Isomaltidex, Palatinit, Palatinit C, Palatinit GS, Palatinit GSP, Palatinit M, Palatinit PM, Palatinit PN, Palatinit PNM 2, Palatinit PNP, Palatinitol, Palatinose PNP
IUPAC Name (3R,4R,5R)-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,4,5-pentol
SMILES C(C1C(C(C(C(O1)OCC(C(C(C(CO)O)O)O)O)O)O)O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H24O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4-21H,1-3H2/t4?,5-,6-,7-,8-,9-,10+,11-,12+/m1/s1
Formula Weight 344.1319
Percent Purity >95%
Chemical Name or Material Isomalt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.