missing translation for 'onlineSavingsMsg'
Learn More

Isomannide Dinitrate, TRC

Product Code. 30005497
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005497 1 g 1g
30003406 100 mg 100mg
30003895 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005497 Supplier Toronto Research Chemicals Supplier No. TRCI8213351G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-43-9
Molecular Formula C6 H8 N2 O8
Molecular Weight (g/mol) 236.14
Synonym Isomannit Dinitrate
IUPAC Name [(3R,3aS,6R,6aS)-6-nitrooxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3-yl] nitrate
SMILES [O-][N+](=O)O[C@@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H8N2O8/c9-7(10)15-3-1-13-6-4(16-8(11)12)2-14-5(3)6/h3-6H,1-2H2/t3-,4-,5-,6-/m1/s1
Formula Weight 236.0281
Chemical Name or Material Isomannide Dinitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.