missing translation for 'onlineSavingsMsg'
Learn More

Isoniazid, TRC

Product Code. 30137154
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137154 1 g 1g
17953255 50 mg 50mg
17993355 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137154 Supplier Toronto Research Chemicals Supplier No. TRCI8214501G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6 H7 N3 O
Molecular Weight (g/mol) 137.14
Synonym Isoniazid, 4-Pyridinecarboxylic acid, hydrazide (9CI, ACI), Isonicotinic acid hydrazide (8CI), 4-(Hydrazinocarbonyl)pyridine, 4-Pyridinecarbonylhydrazine, 4-Pyridinecarboxylic hydrazide, 4-Pyridylcarbonylhydrazide, 5015 R.P., [(4-Pyridinylcarbonyl)oxy]hydrazine, Armazid, Armazide, Atcotibine, Cedin, Cotinazin, Dianicotyl, Dinacrin, Ditubin, Eralon, Ertuban, Eutizon, FSR 3, GINK, HIA, Hidranizil, Hidrasonil, Hycozid, Hydrazid, Hyzyd, Ipcazide, Isidrina, Isobicina, Isocaldin, Isocid, Isocotin, Isokin, Isolyn, Isonex, Isoniazid SA, Isoniazide, Isonicid, Isonico, Isonicotan, Isonicotinic acid hydrazyde, Isonicotinic hydrazide, Isonicotinohydrazide, Isonicotinoyl hydrazide, Isonicotinoylhydrazine, Isonidrin, Isonilex, Isonindon, Isonizide, Isotebezid, Isozide, Isozin, Isozyd, Lafepe, Laniazid, Mayambutol, MeSH ID: D007538, Mybasan, Neoteben, Neoxin, Neumandin, Nevin, Niadrin, Nicetal, Nicizina, Niconyl, Nicotibina, Nicotisan, Nicozide, Nidaton, Nidrazid, Nikozid, Niplen, Nitadon, NSC 9659, Nydrazid, Nyscozid, Pelazid, Preparation 6424, Pycazide, Pyricidin, Pyridine-4-carbohydrazide, Pyrizidin, Raumanon, Retozide, Rimicid, Rimifon, Rimiphone, Rimitsid, Robisellin, RU-EF-Tb, Sauterazid, Solonex, T.B. Razide, TB-Vis, Tebecid, Teebaconin, Tekazin, Tibinide, Tibizide, Tisin, Tisiodrazida, Tizide, Tubazid, Tubazide, Tubeco, Tubicon, Tubilysin, Tubomel, Tyvid, Unicozyde, Vazadrine, Vederon, Zidafimia, Zinadon, Zonazide
IUPAC Name pyridine-4-carbohydrazide
SMILES NNC(=O)c1ccncc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H7N3O/c7-9-6(10)5-1-3-8-4-2-5/h1-4H,7H2,(H,9,10)
Formula Weight 137.0589
Percent Purity >95%
Chemical Name or Material Isoniazid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.