missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic acid, 99%

Product Code. 11429266
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429266 2500 g 2500g
11409266 100 g 100g
11419266 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429266 Supplier Thermo Scientific Alfa Aesar Supplier No. A18109.0E
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55-22-1 | C6H5NO2 | 123.111 g/mol

It acts as an useful isomer of nicotinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as an useful isomer of nicotinic acid.

Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.

Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-22-1
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006429
InChI Key TWBYWOBDOCUKOW-UHFFFAOYSA-N
Synonym isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
PubChem CID 5922
ChEBI CHEBI:6032
IUPAC Name pyridine-4-carboxylic acid
SMILES C1=CN=CC=C1C(=O)O

Specifications

Melting Point ∼310°C (sublimation)
pH 3.0 to 4.0
Boiling Point 260°C (15 mmHg)
Quantity 2500 g
Beilstein 109599
Merck Index 14,5187
Solubility Information Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Isonicotinic acid
compliance-icons
Product Identifier
  • Isonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Isonicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.