missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic acid hydrazide, 98+%

Product Code. 11428870
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428870 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428870 Supplier Thermo Scientific Alfa Aesar Supplier No. A10583.36
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 54-85-3 | C6H7N3O | 137.142 g/mol

Used for fluorescent HPLC detection of delta 4-3-ketosteroids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.142
MDL Number MFCD00006426
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
Synonym isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
PubChem CID 3767
ChEBI CHEBI:6030
IUPAC Name pyridine-4-carbohydrazide
SMILES C1=CN=CC=C1C(=O)NN

Specifications

Melting Point 169°C to 174°C
Quantity 500 g
Beilstein 119374
Sensitivity Air Sensitive
Merck Index 14,5186
Formula Weight 137.14
Percent Purity ≥98%
Chemical Name or Material Isonicotinic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.