missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl 3-methylbutanoate, 98%, BLD Pharm™

Product Code. 31426868
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31426868 100 g
31418505 25 g
31419183 500 g
31425861 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31426868 Supplier BLD Pharm Supplier No. BD134986100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 659-70-1
Molecular Formula C10H20O2
MDL Number MFCD00048369
Quantity 100 g
SMILES CC(C)CC(OCCC(C)C)=O
Molecular Weight (g/mol) 172.26
Percent Purity 98%
Chemical Name or Material Isopentyl 3-methylbutanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.